missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total FGFR4 DuoSet IC ELISA
Click to view available options
Packaging Quantity:
2 Plates
5 Plates
Unit Size:
1 set
Description
The Human Total FGF R4 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human FGFR4.
Specifications
Specifications
| Product Type | ELISA Development Kit for Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2264 |
| Synonym | CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGF R4, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase |
| Packaging Quantity | 5 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction