missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total MMP-1 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
The Human Total MMP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MMP-1.
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4312 |
| Host Species | Human |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 |
| Target | MMP-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction