missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRA-1-81 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB8495
This item is not returnable.
View return policy
Description
TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| TRA-1-81 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PODXL | |
| Human embryonal carcinoma cell line 2102Ep | |
| 100 μg | |
| Primary | |
| Detects human TRA-1-81 in flow cytometry. | |
| Human | |
| Supernatant |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| TRA-1-81 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| gp200, MGC138240, PC, PCLP, PODXL, TRA181 | |
| Mouse | |
| IgM-specific Affinity-purified from hybridoma culture supernatant | |
| RUO | |
| 5420 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction