missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAF-2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3277
This item is not returnable.
View return policy
Description
TRAF-2 Polyclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| TRAF-2 | |
| Polyclonal | |
| Unconjugated | |
| Q12933 | |
| TRAF2 | |
| E. coli-derived recombinant human TRAF-2 Met1-Leu501 Accession # Q12933 | |
| 100 μg | |
| Primary | |
| Detects human TRAF-2 in Western blots. In Western blots, less than 1% cross-reacivity with recombinant human TRAF-1, -3, -4, -5, or -6 is observed. | |
| Human | |
| IgG |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Knockout Validated | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAF2, TRAP3, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7186 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction