missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAF-6 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3284U-100UG
This item is not returnable.
View return policy
Description
TRAF-6 Monoclonal specifically detects TRAF-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TRAF-6 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6, TRAF6 | |
| E.coli-derived recombinant human TRAF-6 Met1-Val522 Accession # Q9Y4K3 | |
| 100 μg | |
| Primary | |
| Detects human TRAF-6 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 326019 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y4K3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7189 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction