missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAIL R3/TNFRSF10C Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB630
This item is not returnable.
View return policy
Description
TRAILR3/TNFRSF10C Monoclonal specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Neutralization.
Specifications
| TRAILR3/TNFRSF10C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAILR3, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 8794 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 90903 | |
| Neutralization 0.04-0.08 ug/mL | |
| O14798 | |
| TNFRSF10C | |
| Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C Ala26-Ala221 Accession # O14798 | |
| 500 μg | |
| Primary | |
| Detects human TRAIL R3/TNFRSF10C in direct ELISAs and Western blots. In direct ELISAs, approximately 20%-40% cross-reactivity with recombinant human (rh) TRAIL R1 is observed and no cross-reactivity with rhTRAIL R2 or rhTRAIL R4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction