missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAILR1/TNFRSF10A Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems BAF347
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Polyclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Western Blot.
Specifications
| TRAILR1/TNFRSF10A | |
| Polyclonal | |
| Biotin | |
| AAC51226 | |
| TNFRSF10A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TRAIL R1/TNFRSF10A Ala24-Asn239 Accession # AAC51226 | |
| 50 μg | |
| Primary | |
| Detects human TRAIL R1/TNFRSF10A in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) TRAIL R2 is observed and no cross-reactivity with rhTRAIL R3 and rhTRAIL R4 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TNFRSF10A, TRAIL R1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8797 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction