missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM-1 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems FAB1278P
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
| TREM1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54210 | |
| Human | |
| Purified |
| Flow Cytometry, Immunoassay | |
| 193015 | |
| Flow Cytometry 10 uL/10^6 cells, Knockout Validated | |
| Q9NP99 | |
| TREM1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TREM-1 Ala21-Asn205 Accession # Q9NP99 | |
| 100 Tests | |
| Primary | |
| Detects human TREM-1 in ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction