Learn More
Human TRIF/TICAM1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF6216
This item is not returnable.
View return policy
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| TRIF/TICAM1 | |
| Polyclonal | |
| Unconjugated | |
| Q8IUC6 | |
| TICAM1 | |
| E. coli-derived recombinant human TRIF/TICAM1 Lys29-Ala204 Accession # Q8IUC6 | |
| 100 μg | |
| Primary | |
| Detects human TRIF/TICAM1 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM1, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 148022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |