missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRIM8 (aa 205-285) Control Fragment Recombinant Protein

Product Code. 30206327
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206327

Brand: Invitrogen™ RP93628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54823 (PA5-54823. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZR9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 81603
Name Human TRIM8 (aa 205-285) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA408830; BB006755; E3 ubiquitin-protein ligase TRIM8; GERP; glioblastoma expressed ring finger protein; glioblastoma-expressed RING finger protein; Probable E3 ubiquitin-protein ligase TRIM8; ring finger protein 27; RING-type E3 ubiquitin transferase TRIM8; Rnf27; TRIM8; tripartite motif containing 8; tripartite motif protein 8; tripartite motif protein TRIM8; tripartite motif-containing 8; tripartite motif-containing protein 8; tripartite motif-containing protein 8; probable E3 ubiquitin-protein ligase TRIM8
Common Name TRIM8
Gene Symbol TRIM8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QDIEDQLYKLESDKRLVEEKVNQLKEEVRLQYEKLHQLLDEDLRQTVEVLDKAQAKFCSENAAQALHLGERMQEAKKLLGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.