missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TROY/TNFRSF19 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1548
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Polyclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Western Blot.
Specifications
| TROY/TNFRSF19 | |
| Polyclonal | |
| Biotin | |
| AAF71828 | |
| TNFRSF19 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TROY/TNFRSF19 Glu30-Leu170 Accession # AAF71828 | |
| 50 μg | |
| Primary | |
| Detects human TROY/TNFRSF19 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhOPG, rhTNF RI, and rhTNF RII is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55504 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction