Learn More
Human Trypsin Pan Specific (PRSS1/2/3) Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems BAF3586
Product Code. 18101471
  • € 560.00 / 50µg
Description
Trypsin Pan Specific/PRSS1/2/3 Polyclonal specifically detects Trypsin Pan Specific/PRSS1/2/3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Trypsin Pan Specific/PRSS1/2/3
Polyclonal
Biotin
P07478
Sheep
Affinity Purified
RUO
5645
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
Mesotrypsin, MTG, PRSS1, PRSS2, PRSS3, PRSS4, PTRYI, Tesp4, TRY2, TRY4, TRY8, Trygn16, TRYP2, Trypsin 4, Trypsinogen 1, Trypsinogen 15, Trypsinogen 20
Mouse myeloma cell line NS0-derived recombinant human Trypsin 2 (R&D Systems, Catalog # 3586-SE) Ala16-Ser247 Accession # P07478
50 μg
Primary
Detects human Trypsin 1, human Trypsin 2 and human Trypsin 3 in Western blots.