Learn More
Human Trypsin Pan Specific (PRSS1/2/3) Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF3586
This item is not returnable.
View return policy
Description
Trypsin Pan Specific/PRSS1/2/3 Polyclonal specifically detects Trypsin Pan Specific/PRSS1/2/3 in Human samples. It is validated for Western Blot.
Specifications
| Trypsin Pan Specific/PRSS1/2/3 | |
| Polyclonal | |
| Biotin | |
| P07478 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5645 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Mesotrypsin, MTG, PRSS1, PRSS2, PRSS3, PRSS4, PTRYI, Tesp4, TRY2, TRY4, TRY8, Trygn16, TRYP2, Trypsin 4, Trypsinogen 1, Trypsinogen 15, Trypsinogen 20 | |
| Mouse myeloma cell line NS0-derived recombinant human Trypsin 2 (R&D Systems, Catalog # 3586-SE) Ala16-Ser247 Accession # P07478 | |
| 50 μg | |
| Primary | |
| Detects human Trypsin 1, human Trypsin 2 and human Trypsin 3 in Western blots. | |
| Human | |
| IgG |