missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tryptase beta-2/TPSB2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB37961
341.70 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Tryptase beta-2/TPSB2 Monoclonal specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
Specifications
| Tryptase beta-2/TPSB2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.4 ug/mL | |
| AAD13876 | |
| TPSB2 | |
| Mouse myeloma cell line NS0-derived recombinant human Tryptase beta-2/TPSB2 Ile31-Pro275 Accession # AAD13876 | |
| 100 μg | |
| Primary | |
| Detects human Tryptase beta-2/TPSB2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TPS1, rhTPSG1, recombinant mouse (rm) TPS5, rmMCPT1, rmMCPT6, rmMCPT7, or rmMCPT11 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Neutralization | |
| 349414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2, TPS2tryptase beta-2, TPSB2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase beta2, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC, Tryptase-C | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64499 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction