missing translation for 'onlineSavingsMsg'
Learn More

Human Tryptase beta-2/TPSB2 Antibody, R&D Systems™

Product Code. 18711239
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18711239 25 μg 25µg
18150621 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18711239 Supplier R&D Systems Supplier No. MAB37961SP
  • ONLINE EXCLUSIVE PRICE
    € 213.78
    € 190.05 / 25µg
    Save € 23.73
    11% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tryptase beta-2/TPSB2 Monoclonal specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for Immunoprecipitation∣Neutralization∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tryptase beta-2/TPSB2
Applications Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone 349414
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAD13876
Gene Alias EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2, TPS2tryptase beta-2, TPSB2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase beta2, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC, Tryptase-C
Gene Symbols TPSB2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase beta-2/TPSB2 Ile31-Pro275 Accession # AAD13876
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64499
Test Specificity Detects human Tryptase beta-2/TPSB2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TPS1, rhTPSG1, recombinant mouse (rm) TPS5, rmMCPT1, rmMCPT6, rmMCPT7, or rmMCPT11 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.