missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSGA2 (aa 149-224) Control Fragment Recombinant Protein

Product Code. 30208880
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208880

Brand: Invitrogen™ RP91776

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53601 (PA5-53601. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a male meiotic metaphase chromosome-associated acidic protein. This gene is expressed in tissues with motile cilia or flagella, including the trachea, lungs, airway brushings, and testes. Mutations in this gene result in primary ciliary dyskinesia-24. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2014].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WYR4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 89765
Name Human TSGA2 (aa 149-224) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ABCB2; cancer/testis antigen 79; CT79; h-meichroacidin; Male meiotic metaphase chromosome-associated acidic protein; MCA; Meichroacidin; MGC126568; MGC141927; PSF1; radial spoke head 1 homolog; radial spoke head 1 homolog (Chlamydomonas); RING4; RSP44; Rsph1; RSPH10A; testes specific gene A2 homolog; testis specific A2 homolog; testis specific gene A2; testis-specific gene A2 protein; TSA2; TSGA2; TSGA2 epididymal isoform
Common Name TSGA2
Gene Symbol RSPH1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TAELIHLNHRYQGKFLNKNPVGPGKYVFDVGCEQHGEYRLTDMERGEEEEEEELVTVVPKWKATQITELALWTPTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.