missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSLPR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF981
This item is not returnable.
View return policy
Description
TSLPR/CRLF2 Polyclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| TSLPR/CRLF2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| CRL2, CRL2cytokine receptor CRL2 precusor, CRLF2, CRLF2Y, CRLM-2, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, PCOR1, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP R, TSLP receptor, TSLPR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64109 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.2-1 μg/mL, Immunohistochemistry 5-15 μg/mL, Neutralization 1-5 μg/mL, Immunocytochemistry 5-25 μg/mL | |
| Q9HC73 | |
| CRLF2 | |
| Mouse myeloma cell line NS0-derived recombinant human TSLP R Val30-Phe232 Accession # Q9HC73 | |
| 100 μg | |
| Primary | |
| Detects human TSLP R in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction