missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSP50 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2430
This item is not returnable.
View return policy
Description
TSP50 Polyclonal specifically detects TSP50 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TSP50 | |
| Polyclonal | |
| Unconjugated | |
| Q9UI38 | |
| PRSS50 | |
| Mouse myeloma cell line NS0-derived recombinant human TSP50 Ala40-Ala363 Accession # Q9UI38 | |
| 100 μg | |
| Primary | |
| Detects human TSP50 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse TSP50 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunoprecipitation 25 ug/mL | |
| Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29122 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction