missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSPAN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7876
This item is not returnable.
View return policy
Description
TSPAN2 Monoclonal specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| TSPAN2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| O60636 | |
| TSPAN2 | |
| NS0 mouse myeloma cell line transfected with human TSPAN2 Met1-Ile221 Accession # O60636 | |
| 100 μg | |
| Primary | |
| Detects human TSPAN2 in ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 822509 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, Tspan-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10100 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction