missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Twist-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6230RG-100UG
This item is not returnable.
View return policy
Description
Twist-1 Monoclonal specifically detects Twist-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Twist-1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twist, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, Twist1, twist-related protein 1 | |
| E.coli-derived recombinant human Twist-1 Met1-His202 Accession # Q15672 | |
| 100 μg | |
| 7291 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 927403R | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/10^6 cells | |
| Q15672 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Twist-1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction