missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Twist-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6230-SP
This item is not returnable.
View return policy
Description
Twist-1 Polyclonal specifically detects Twist-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Twist-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twist, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, Twist1, twist-related protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7291 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| Q15672 | |
| TWIST1 | |
| E. coli-derived recombinant human Twist-1 Met1-His202 Accession # Q15672 | |
| 25 μg | |
| Primary | |
| Detects human Twist-1 in direct ELISAs. In direct ELISAs, less than 16% cross-reactivity with recombinant human Twist-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction