missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TXN (NP_003320, 1 a.a. - 105 a.a.) Full-length Recombinant Protein

Product Code. 16171350
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16171350

Brand: Abnova™ P3454.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Thioredoxin is a 12-kD oxidoreductase enzyme containing a dithiol-disulfide active site. It is ubiquitous and found in many organisms from plants and bacteria to mammals. Multiple in vitro substrates for thioredoxin have been identified, including ribonuclease, choriogonadotropins, coagulation factors, glucocorticoid receptor, and insulin. Reduction of insulin is classically used as an activity test.[supplied by OMIM]

Sequence: MVKQIESKTAFQEALDAAGDKLVVVDFSATWCGPCKMIKPFFHSLSEKYSNVIFLEVDVDDCQDVASECEVKCMPTFQFFKKGQKVGEFSGANKEKLEATINELV

Specifications

Accession Number NP_003320
Concentration 1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 7295
Molecular Weight (g/mol) 11.7kDa
Name TXN (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Immunogen MVKQIESKTAFQEALDAAGDKLVVVDFSATWCGPCKMIKPFFHSLSEKYSNVIFLEVDVDDCQDVASECEVKCMPTFQFFKKGQKVGEFSGANKEKLEATINELV
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686B1993/MGC61975/TRX/TRX1
Common Name TXN
Gene Symbol TXN
Biological Activity Specific activity is 7-10 A650/min/mg, obtained by measuring the increase of insulin precipitation in absorbance at 650nm resulting from the reduction of insulin. Please refer to our activity assay protocol.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.