missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tyro3/Dtk Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB859T-100UG
This item is not returnable.
View return policy
Description
Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Dtk/TYRO3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tyro3/Dtk Ala41-Ser428 (predicted) Accession # Q06418 | |
| Primary | |
| Detects human Tyro3/Dtk in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity or interferance with recombinant mouse Tyro3/Dtk, recombinant human (rh) Axl, or rhMer is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 96201 | |
| Flow Cytometry 0.25-1μg/106 cells | |
| Q06418 | |
| Mouse | |
| 100 μg | |
| 7301 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction