missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UACA (aa 870-972) Control Fragment Recombinant Protein

Product Code. 30208507
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208507

Brand: Invitrogen™ RP106043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65361 (PA5-65361. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZF9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55075
Name Human UACA (aa 870-972) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700059D02Rik; beta-actin binding protein betaCAP73; beta-actin-binding protein; BetaCAP73; Kiaa1561; nuclear membrane binding protein; nuclear membrane binding protein NUCLING; nuclear membrane-binding protein; Nucling; RGD1560011; Uaca; uveal autoantigen; uveal autoantigen with coiled coil domains and ankyrin repeats; uveal autoantigen with coiled-coil domains and ankyrin repeats; Uveal autoantigen with coiled-coil domains and ankyrin repeats protein
Common Name UACA
Gene Symbol UACA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MTLNDTLAKTNRELLDVKKKFEDINQEFVKIKDKNEILKRNLENTQNQIKAEYISLAEHEAKMSSLSQSMRKVQDSNAEILANYRKGQEEIVTLHAEIKAQKK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.