missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Ubiquilin 1 (aa 348-376) Control Fragment Recombinant Protein

Product Code. 30203981
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203981

Brand: Invitrogen™ RP104987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer's and Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UMX0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 29979
Name Human Ubiquilin 1 (aa 348-376) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110046H03Rik; 1810030E05Rik; AU019746; C77538; D13Ertd372e; Da41; Dsk2; hPLIC-1; I79_020705; Plic1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; ubiquilin 1; ubiquilin-1; UBQLN1; UBQN; XDRP1
Common Name Ubiquilin 1
Gene Symbol Ubqln1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence STASGTSGQSTTAPNLVPGVGASMFNTPG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.