missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-1 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1380T-100UG
This item is not returnable.
View return policy
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| ULBP-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human ULBP-1 cell transfectants. It does not stain cells transfected with ULBP-2 or ULBP-3. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 170818 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ULBP1 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-1 | |
| RUO | |
| 80329 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction