missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-1 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems FAB1380P
This item is not returnable.
View return policy
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| ULBP-1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| ULBP1 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-1 | |
| 100 Tests | |
| Primary | |
| Stains human ULBP-1 cell transfectants. It does not stain cells transfected with ULBP-2 or ULBP-3. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 170818 | |
| Flow Cytometry 10 uL/10^6 cells | |
| ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 80329 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction