missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-4/RAET1E Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6285S-100UG
This item is not returnable.
View return policy
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry.
Specifications
| ULBP-4/RAET1E | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8TD07 | |
| RAET1E | |
| Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E Gly30-Asp225 Accession # Q8TD07 | |
| RUO | |
| 135250 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 709116 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| bA350J20.7, LETAL, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E, RAET1E2, retinoic acid early transcript 1ERL-4, RL-4, ULBP4, ULBP4MGC125308 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ULBP-4/RAET1E in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) ULBP-2 is observed and no cross-reactivity with rhULBP-1 or rhULBP-3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction