missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UQCC1 (aa 166-292) Control Fragment Recombinant Protein

Product Code. 30206605
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206605

Brand: Invitrogen™ RP90031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57087 (PA5-57087. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NVA1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55245
Name Human UQCC1 (aa 166-292) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310079L17Rik; 2410003P15Rik; 3110038N19Rik; basic FGF repressed, Zic binding protein; basic FGF repressed, zinc binding protein; basic FGF-repressed Zic-binding protein; bFGF-repressed Zic-binding protein; BFZB; Bfzp; BZFB; C20orf44; Cbp3; cytochrome B protein synthesis 3 homolog; FLJ10850; mbFZb; ubiquinol-cytochrome c reductase complex assembly factor 1; ubiquinol-cytochrome c reductase complex chaperone; ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog; ubiquinol-cytochrome c reductase complex chaperone, CBP3 homolog; ubiquinol-cytochrome-c reductase complex assembly factor 1; UQCC; UQCC1
Common Name UQCC1
Gene Symbol UQCC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RSGKYMCRIIVHFMWEDVQQRGRVMGVNPYILKKNMILMTNHFYAAILGYDEGILSDDHGLAAALWRTFFNRKCEDPRHLELLVEYVRKQIQYLDSMNGEDLLLTGEVSWRPLVEKNPQSILKPHSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.