missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human USAG1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7034-SP
This item is not returnable.
View return policy
Description
USAG1/SOSTDC1 Polyclonal specifically detects USAG1/SOSTDC1 in Human samples. It is validated for Western Blot.
Specifications
| USAG1/SOSTDC1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDA019, cystine-knot containing secreted protein, DKFZp564D206, Ectodermal BMP inhibitor, ECTODIN, sclerostin domain containing 1, sclerostin domain-containing protein 1, SOSTDC1, USAG-1, USAG1uterine sensitization-associated protein-1, Uterine sensitization-associated gene 1 protein, WISE | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 25928 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6X4U4 | |
| SOSTDC1 | |
| E. coli-derived recombinant human USAG1 Phe24-Ser206 Accession # Q6X4U4 | |
| 25 μg | |
| Primary | |
| Detects human USAG1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction