missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VAMP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6117-SP
This item is not returnable.
View return policy
Description
VAMP-7 Monoclonal specifically detects VAMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VAMP-7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P51809 | |
| VAMP7 | |
| E. coli-derived recombinant human VAMP-7 Ala2-Met140 Accession # P51809 | |
| 25 μg | |
| Primary | |
| Detects human VAMP-7 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 549115 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ54296, SYBL1, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, TIVAMP, TI-VAMP, VAMP7, VAMP-7FLJ53045, vesicle-associated membrane protein 7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6845 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction