missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VAP-B Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC58551R-100UG
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
VAP-B Monoclonal specifically detects VAP-B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifikationer
| VAP-B | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O95292 | |
| VAPB | |
| E. coli-derived recombinant human VAP-B Ala2-Pro132 Accession # O95292 | |
| RUO | |
| 9217 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 736904 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ALS8, ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAMP-C, VAPB, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human VAP-B in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human VAP-A is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only (RUO)
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering