missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VDR/NR1I1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems PP-H4537-00
This item is not returnable.
View return policy
Description
VDR/NR1I1/Vitamin D Receptor Monoclonal specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.
Specifications
| VDR/NR1I1/Vitamin D Receptor | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| VDR | |
| Recombinant human VDR/NR1I1 aa 91-210 | |
| 100 μg | |
| Primary | |
| This antibody specifically recognizes human VDR and cross reacts with mouse and rat VDR. Not yet tested in other species. | |
| This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA | |
| H4537 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA | |
| NR1I1, NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7421 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction