missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human VEGF-A Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human VEGF-A.
Merk: Invitrogen™ BMS277-2TENCE
Overige gegevens : Gewicht : 0.23750kg
Inclusief: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human VEGF-A
Biotin-Conjugate anti-human VEGF-A polyclonal antibody
Streptavidin-HRP
Human VEGF-A Standard lyophilized, 2ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Beschrijving
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specificatie
Biotin | |
10 x 96 tests | |
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin) | |
Human | |
Vascular endothelial growth factor A, VPF |
ELISA | |
7.9pg/mL | |
50μL | |
VEGF-A |
For Research Use Only