missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2931G
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| VEGF | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| VEGFA | |
| E. coli-derived recombinant human VEGF | |
| 100 Tests | |
| Primary | |
| Detects human VEGF in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 23410 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7422 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction