missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human VEGF Quantikine QuicKit Control Group 285
Click to view available options
Quantity:
1 Ea.
Unit Size:
1 set
Specifications
Specifications
| For Use With (Application) | Control |
| Gene ID (Entrez) | 7422 |
| Quantity | 1 Ea. |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor |
| Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction