missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WIF-1 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems BAM1341
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| WIF-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| AAD25402 | |
| WIF1 | |
| Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402 | |
| 250 μg | |
| Primary | |
| Detects human WIF-1 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with recombinant mouse WIF-1 or recombinant human Wnt-7a is observed. In Western blots, 50% cross-reactivity with recombinant mouse WIF-1 is observed. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 133016 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| WIF1, WIF-1, WNT inhibitory factor 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11197 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction