missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human WISP-1/CCN4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1627
This item is not returnable.
View return policy
Description
WISP-1/CCN4 Polyclonal specifically detects WISP-1/CCN4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| WISP-1/CCN4 | |
| Polyclonal | |
| Unconjugated | |
| O95388 | |
| WISP1 | |
| Mouse myeloma cell line NS0-derived recombinant human WISP-1/CCN4 Thr23-Asn367 Accession # O95388 | |
| 100 μg | |
| Primary | |
| Detects human WISP-1/CCN4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse WISP-1 is observed and less than 1% cross-reactivity with recombiant human (rh) CTGF and rhNOV is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8840 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction