Learn More
R&D Systems™ Human XL Cytokine Luminex™ Performance Assay 46-plex Fixed Panel
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |