Learn More
- Home
- Products
- Immunoassay Reagents and Kits
- Singleplex and Multiplex Bead Based Immunoassays
- R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
R&D Systems™ Human XL Cytokine Luminex™ Performance Panel Premixed Kit
Product Code. 30000840
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
46 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
Each
45 product options available for selection
| Product Code. | Quantity | unitSize |
|---|---|---|
| 30000840 | 14 Plex | Each |
| 30001034 | 23 Plex | Each |
| 30001033 | 24 Plex | Each |
| 30001030 | 28 Plex | Each |
| 30001019 | 39 Plex | Each |
| 30001017 | 17 Plex | Each |
| 30001001 | 5 Plex | Each |
| 18722587 | 31 Plex | Each |
| 30000992 | 41 Plex | Each |
| 30000983 | 18 Plex | Each |
| 30000979 | 43 Plex | Each |
| 30000978 | 11 Plex | Each |
| 30000957 | 27 Plex | Each |
| 30000955 | 37 Plex | Each |
| 30000937 | 45 Plex | Each |
| 18793377 | 42 Plex | Each |
| 30000922 | 44 Plex | Each |
| 30000920 | 6 Plex | Each |
| 18754277 | 3 Plex | Each |
| 30000908 | 30 Plex | Each |
| 30000906 | 38 Plex | Each |
| 30000899 | 15 Plex | Each |
| 18732627 | 10 Plex | Each |
| 18702377 | 33 Plex | Each |
| 18704447 | 8 Plex | Each |
| 18782497 | 36 Plex | Each |
| 30000858 | 16 Plex | Each |
| 18700617 | 7 Plex | Each |
| 30000854 | 19 Plex | Each |
| 30000841 | 20 Plex | Each |
| 30000832 | 4 Plex | Each |
| 30000828 | 35 Plex | Each |
| 18792687 | 9 Plex | Each |
| 30000813 | 2 Plex | Each |
| 30000796 | 26 Plex | Each |
| 30000786 | 46 Plex | Each |
| 30000770 | 29 Plex | Each |
| 30000761 | 25 Plex | Each |
| 30000757 | 12 Plex | Each |
| 30000755 | 22 Plex | Each |
| 30000745 | 34 Plex | Each |
| 30000743 | 32 Plex | Each |
| 30000742 | 40 Plex | Each |
| 30000738 | 21 Plex | Each |
| 18772167 | 13 Plex | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
Specifications
| Quantity | 14 Plex |
| Species | Human |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |