missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ZnT-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7936-SP
This item is not returnable.
View return policy
Description
ZnT-8/SLC30A8 Monoclonal specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| ZnT-8/SLC30A8 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q8IWU4 | |
| SLC30A8 | |
| E. coli-derived recombinant human ZnT-8 Lys268-Pro359 Accession # Q8IWU4 | |
| 25 μg | |
| Primary | |
| Detects human ZnT-8 in ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 815039 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SLC30A8, solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZnT8, ZNT8ZnT-8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 169026 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction