Learn More
Proteintech™ HumanKine™ Recombinant Human BMP-2 Protein
Product Code. 16881326 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. 16881326
Brand: Proteintech™ HZ1128100UG
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Gene ID (Entrez) 650
Molecular Weight (g/mol) 28 kDa, homodimer, glycosylated
Quantity 100 μg
Species Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 4 mM HCl containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Content And Storage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Activity Typically ≤ 60 ng/mL EC50
Expression System Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Purity or Quality Grade > 95%