Learn More
Rotronic™ Humidity standards
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
€ 108.00 - € 126.00
Specifications
Certificate of Analysis | SCS traceable calibration certificate, textile pad, calibration instruction |
---|---|
For Use With (Equipment) | Rotronic Instrument |
For Use With (Application) | On-site calibration and adjustment of rotronic probe |
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
11980745
|
Rotronic™
EA00-SCS |
0.005 |
€ 108.00
Pack of 5 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11930665
|
Rotronic™
EA80SCS |
0.8 |
€ 126.00
Pack of 5 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11960655
|
Rotronic™
EA10SCS |
0.1 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
- Ampoules contain unsaturated salt solutions
- Simple and safe on-site calibration method
- Unlimited storage life
Specifications
SCS traceable calibration certificate, textile pad, calibration instruction | |
Rotronic Instrument | |
On-site calibration and adjustment of rotronic probe |