missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuR/ELAVL1 Antibody (OTI14F4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70635AF594
This item is not returnable.
View return policy
Description
HuR/ELAVL1 Monoclonal antibody specifically detects HuR/ELAVL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HuR/ELAVL1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG | |
| Full length recombinant protein of human HuR/ELAVL1 (NP_001410) produced in HEK293T cells. | |
| 0.1 mL | |
| Vision | |
| 1994 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI14F4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction