missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | HUS1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229821
|
Novus Biologicals
NBP3-38146-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227265
|
Novus Biologicals
NBP3-38146-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HUS1 Polyclonal antibody specifically detects HUS1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| HUS1 | |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 3364 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| checkpoint protein HUS1, hHUS1, HUS1 (S. pombe) checkpoint homolog, HUS1 checkpoint homolog (S. pombe), hus1+-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HUS1 (NP_004498.1).,, Sequence:, MKFRAKIVDGACLNHFTRISNMIAKLAKTCTLRISPDKLNFILCDKLANGGVSMWCELEQENFFNEFQMEGVSAENNEIYLELTSENLSRALKTAQNARALKIKLTNKHFPCLTVSVELLSMSSSSRIVTHDIPIKVIPRKLWKDLQEPVVPDPDVSIYLPVLKTMKSVVEKMKNISNHLVIEANLDGELNLKIETELVCVTTHFKDLGNPPLASESTHEDRNVEHMAEVHIDIRKLLQFLAGQQVNPTKALCNIVNNKMVHFDLLHEDVSLQYFIPALS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title