Learn More
Hamilton™ HVCX Large Body Ceramic Valves
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
4-5 Model
8-5 Model
For Use With:
PSD/2, 3 Syringe Drive Module
PSD/4, 8 Syringe Drive Module
missing translation for 'unitSize'
Each
Description
- Rigid ceramic fluid path is resistant to scratching from salt crystals
- Provides superior life and performance
- Exceptionally easy to service and modify
- Maximizes flexibility and value
- Ideal for low pressure applications
Specifications
Specifications
| Description | 4-5 Model, 4 Ports, Stainless Steel Housing |
| For Use With | PSD/2, 3 Syringe Drive Module |
| Material | Stainless Steel |
| Model | 4-5 Model |
| Type | Ceramic Large Body Valve |