missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HVEM/TNFRSF14 Antibody (2742B), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB3563W
This item is not returnable.
View return policy
Description
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Flow Cytometry, Neutralization
Specifications
| HVEM/TNFRSF14 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization | |
| 2742B | |
| Flow Cytometry, Neutralization | |
| CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
| Human embryonic kidney cell HEK293-derived human HVEM/TNFRSF14, Pro37-Val202, Accession # Q92956 | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology | |
| 8764 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction