missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HVEM/TNFRSF14 Antibody (HMHV-1B18) - Chimeric - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
€ 250.00 - € 705.00
Specifications
| Antigen | HVEM/TNFRSF14 |
|---|---|
| Clone | HMHV-1B18 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627057
|
Novus Biologicals
NBP3-09028-0.2mg |
0.2 mg |
€ 705.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653549
|
Novus Biologicals
NBP3-09028-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization, Functional AssaySpecifications
| HVEM/TNFRSF14 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
| This antibody was raised by immunising Armenian hamsters with mouse HVEM:Fc fusion protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| HMHV-1B18 | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization, Functional Assay | |
| Unconjugated | |
| Armenian Hamster | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology | |
| PBS | |
| 8764 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title