missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HYAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530283
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (88%), Rat (88%), Pig (93%), Sheep (91%), Rhesus Monkey (100%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a weak acid-active hyaluronidase. The encoded protein is similar in structure to other more active hyaluronidases. Hyaluronidases degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan and fragments of hyaluronan are thought to be involved in cell proliferation, migration and differentiation. Although it was previously thought to be a lysosomal hyaluronidase that is active at a pH below 4, the encoded protein is likely a GPI-anchored cell surface protein. This hyaluronidase serves as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5' UTR.
Specifications
| HYAL2 | |
| Polyclonal | |
| Unconjugated | |
| HYAL2 | |
| AI256841; AU020858; AW555733; HYAL2; Hyal-2; Hyaluronidase; hyaluronidase 2; hyaluronidase-2; hyaluronoglucosaminidase 2; hyaluronoglucosaminidase-2; LUCA2; LuCa-2; lung carcinoma protein 2; lysosomal hyaluronidase; PH20 homolog; PH-20 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15587, 8692 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O35632, Q12891 | |
| HYAL2 | |
| Recombinant protein encompassing a sequence within the center region of human HYAL2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction