Learn More
R&D Systems™ Hyaluronan Quantikine ELISA Kit
Description
The Hyaluronan Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
Specifications
Specifications
| Assay Range | 0.63 - 40ng/mL |
| Assay Sensitivity | 0.2ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
| Specificity | 35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples |
| For Use With (Application) | ELISA |
| Quantity | One 96-well plate |
| Sample Volume | Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |