missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Hyaluronan synthase 1 Antibody (5C9D6), Novus Biologicals™
Mouse Monoclonal Antibody
€ 299.00 - € 524.00
Specifications
| Antigen | Hyaluronan synthase 1 |
|---|---|
| Clone | 5C9D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30034234
|
Novus Biologicals™
NBP3-27161-0.025ml |
0.025 mL |
€ 299.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30034316
|
Novus Biologicals™
NBP3-27161-0.1ml |
0.1 mL |
€ 524.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hyaluronan synthase 1 Monoclonal antibody specifically detects Hyaluronan synthase 1 in Human,Mouse samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| Hyaluronan synthase 1 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells | |
| PBS | |
| 3036 | |
| IgG2b | |
| Protein G purified |
| 5C9D6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1 | |
| Purified recombinant fragment of human Hyaluronan synthase 1 (AA: (74-399)) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title